Learn More
Medchemexpress LLC 1-methyl-4-imidazoleacetic acid hydrochloride | 35454-39-8 | MFCD00036998 | 99.3% | 176.60 | C6H9ClN2O2 | 1 ML

Supplier: Medchemexpress LLC HYW06047010MM/1ML
1-Methyl-4-imidazoleacetic acid hydrochloride is the hydrochloride salt of a stable histamine metabolite used as a reagent and reference standard in biochemical and pharmacological research. It is supplied as a solid and as a 10 mM solution in DMSO (1 mL) for assay and control use.
- Hydrochloride salt of 1-methyl-4-imidazoleacetic acid.
- CAS number 35454-39-8.
- Molecular weight 176.60.
- Purity reported as 98.0% (product listing) and 99.25% (COA); confirm lot COA for exact value.
- Available as solid in multiple mg sizes and as a 10 mM, 1 mL solution in DMSO.
- Store at -20°C sealed and protected from moisture; in solvent store at -80°C up to 6 months.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.