Learn More
Medchemexpress LLC 1-methyl-6-nitro-3,1-benzoxazine-2,4-dione | 4693-01-0 | MFCD00129950 | 95.9% | 222.15 g/mol | C9H6N2O5 | 5 MG

Supplier: Medchemexpress LLC HYW2005455MG
1M6 is a small-molecule reagent for chemical probing and detection of RNA secondary and tertiary structure in laboratory research. Provided as a light yellow to yellow solid, it is intended for research use only and not for human clinical applications.
- Used to chemically detect and probe RNA structure.
- High purity: 95.9%.
- Molecular formula C9H6N2O5; molecular weight 222.15 g/mol.
- Light yellow solid suitable for biochemical assays.
- Available in small-mass quantities for research workflows.
- Recommended storage: powder at -20°C for long-term stability.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.