Learn More
Medchemexpress LLC 1-(methyl-d3)piperazine | 1093380-08-5 | MFCD03428305 | 95.0% | 10 MG

Supplier: Medchemexpress LLC HY78871S10MG
1-(Methyl-d3)piperazine is the deuterium-labeled form of N-methylpiperazine used as a tracer and internal standard in analytical and research applications. It is intended for quantitative analyses by NMR, GC-MS, and LC-MS and is supplied in small laboratory pack sizes for use in isotope-labeling studies.
- Deuterium-labeled N-methylpiperazine (C5H9D3N2)
- Used as a tracer and internal standard for quantitative analysis by NMR, GC-MS, and LC-MS
- Molecular weight 103.18 g/mol
- Physical form: liquid, colorless to yellow
- Available in small pack sizes including 10 MG
- Storage: pure form -20°C; in solvent -80°C (short term)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.