Learn More
Medchemexpress LLC 1-methyl-L-histidine | 332-80-9 | 100.0% | 169.18 g·mol⁻¹ | C7H11N3O2 | 25 MG

Supplier: Medchemexpress LLC 25MGHYW017006
1-Methyl-L-histidine is an amino acid biomarker used as an objective indicator of meat ingestion and exogenous 3-methylhistidine (3MH) intake. Supplied as a white to off-white solid for research use, it has defined solubility and storage properties for laboratory applications.
- High purity (99.97%).
- Molecular formula C7H11N3O2 and molecular weight 169.18 g·mol⁻¹.
- White to off-white solid appearance.
- Water solubility: 125 mg/mL (requires ultrasonic).
- PBS solubility: 100 mg/mL (clear solution; requires ultrasonic).
- Powder storage: -20 °C (3 years), 4 °C (2 years).
- In-solution storage: -80 °C (2 years), -20 °C (1 year).
- Available in small solid pack sizes including 5 mg, 10 mg, and 25 mg.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.