Learn More
Medchemexpress LLC 1-methyl-L-histidine-d3 | 91037-48-8 | 99.4% | 172.20 g/mol | C7H8D3N3O2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYW017006S10MG
Encompass_Preferred
1-Methyl-L-histidine-d3 is the deuterium-labeled isotopologue of 1-methyl-L-histidine used as a tracer and internal standard in analytical workflows. It supports quantitative mass spectrometry and metabolic studies, such as assessing meat ingestion or exogenous 3-methylhistidine intake.
  • Deuterium-labeled internal standard suitable for mass spectrometry and tracer studies.
  • High chemical purity (99.4%) for reliable quantitation.
  • Chemical formula C7H8D3N3O2 and molecular weight 172.20 g/mol.
  • Solid, off-white to light yellow appearance for easy handling.
  • Available in small-package sizes (1-100 mg) to support analytical needs.
  • Stable when stored at recommended conditions, preserving integrity for long-term use.
Catalog No. 50-003-02012