Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-Methylguanidine monohydrochloride | 21770-81-0 | 109.56 | 1 G
SDP

Supplier:  Medchemexpress LLC HYW0050281G

Encompass_Preferred

1-Methylguanidine monohydrochloride is an endogenous metabolite. It is for research use only and not sold to patients.

  • Endogenous metabolite
  • White to off-white solid appearance
  • High purity of 98.0%
  • Molecular weight: 109.56
  • Solid storage: 4°C, sealed, away from moisture
  • In solvent storage: -80°C for 6 months; -20°C for 1 month (sealed, away from moisture)

Catalog No. 50-004-48965


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.