Learn More
Medchemexpress LLC 1-methylhistamine-d3 dihydrochloride | 344299-49-6 | 99.9% | 201.11 g/mol | C6H10D3Cl2N3 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYW053787S5MG
Encompass_Preferred
1-Methylhistamine-d3 dihydrochloride is the deuterium-labeled dihydrochloride salt of 1-methylhistamine, supplied as a high-purity stable isotope standard for analytical applications such as mass spectrometry and metabolite quantification.
  • stable isotope-labeled internal standard for analytical quantification
  • high purity suitable for LC-MS/MS and tracer studies
  • solid dihydrochloride salt form for easy handling and storage
  • available in small sizes for use as reference standards
  • useful for metabolite quantification and tracer experiments
Catalog No. 50-002-09953