Learn More
TARGETMOL CHEMICALS INC 1-METHYLHISTAMINE DIHYDR 50MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T879050MG
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 1-Methylhistamine dihydrochloride is a major metabolite of histamine by histamine N-methyltransferase in the pathway of histidine metabolism. It has been used as a biomarker of histaminergic system activity in the brains of Alzheimer's disease patients as well as those with hypersomnia and other neurological conditions. purity: 99%
Catalog No. 50-257-9045