Learn More
Medchemexpress LLC 1-Methylnicotinamide chloride | 1005-24-9 | 99.9% | 172.61 | 500 MG
Supplier: Medchemexpress LLC HY113527500MG
TRIA-662 (1-Methylnicotinamide chloride) is an endogenous metabolite that exhibits antithrombotic and anti-inflammatory activities. It is intended for research use only. In vitro studies have shown its ability to induce polymerization of G-actin and demonstrate anti-inflammatory activity in the endothelium.
- Endogenous metabolite
- Exhibits antithrombotic activities
- Exhibits anti-inflammatory activities
- Induces polymerization of G-actin in vitro
- Demonstrates anti-inflammatory activity in the endothelium in vitro