Learn More
Medchemexpress LLC 1-Methylnicotinamide-d3 chloride | 1218993-18-0 | 99.4% | 175.63 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY113527S10MG
Encompass_Preferred
TRIA-662-d3 is the deuterium labeled TRIA-662. TRIA-662 (1-Methylnicotinamide chloride) is an endogenous metabolite that shows antithrombotic and anti-inflammatory activities. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium labeled compound
  • Endogenous metabolite
  • Antithrombotic and anti-inflammatory activities
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-36563