Learn More
Medchemexpress LLC 1-methylpiperazine-2,2,3,3,5,5,6,6-d8 | 917358-65-7 | MFCD03428101 | 98.2% | 108.21 g·mol⁻¹ | C5H4D8N2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYW45124210MG
Encompass_Preferred
1-Methylpiperazine-2,2,3,3,5,5,6,6-d8 is a deuterium-labeled analogue of 1-methylpiperazine intended for research use. It serves as a stable isotopic tracer and internal standard for analytical and quantitative workflows, including mass spectrometry and NMR. This product is supplied for laboratory research and is not for human or clinical use.
  • Deuterium-labeled compound for isotopic tracing.
  • High chemical purity suitable for analytical workflows.
  • Compatible with mass spectrometry and NMR analyses.
  • Useful as an internal standard and tracer in quantitative assays.
  • Stable, non-radioactive isotope label for reproducible results.
Catalog No. 50-002-81707