Learn More
Medchemexpress LLC 1-Methylpyrrolidine | 120-94-5 | 99.0% | 500 MG

Supplier: Medchemexpress LLC HY128383500MG
1-Methylpyrrolidine is a methylated pyrrolidine used as a precursor for synthesizing polyfluorinated pyrrolidine derivatives. It is a colorless to light yellow liquid with a density of 0.819 g/cm³. The molecular weight is 85.15 and its chemical formula is C₅H₁₁N. This product is for research use only.
- Purity: 99.02%
- Molecular weight: 85.15
- Chemical formula: C₅H₁₁N
- Appearance: Liquid
- Color: Colorless to light yellow
- SMILES: CN1CCCC1
- Storage for pure form: -20°C for 3 years
- Storage in solvent: -80°C for 6 months, -20°C for 1 month
- In vitro solubility: 100 mg/mL in DMSO
- In vivo solubility: ≥ 2.5 mg/mL in various protocols
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.