Learn More
Medchemexpress LLC 1,N6-Ethenoadenosine | 39007-51-7 | 99.7% | 291.26 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY137600100MG
Encompass_Preferred
1,N6-Ethenoadenosine is a purine nucleoside analogue. These analogues demonstrate broad antitumor activity, particularly against indolent lymphoid malignancies. Their anticancer mechanisms primarily involve the inhibition of DNA synthesis and the induction of apoptosis.
  • Purine nucleoside analogue.
  • Exhibits broad antitumor activity.
  • Targets indolent lymphoid malignancies.
  • Inhibits DNA synthesis.
  • Induces apoptosis.
Catalog No. 50-003-49028