Learn More
Medchemexpress LLC 1-Naphthaleneacetic acid | 86-87-3 | 99.7% | 186.21 | 50 G
SDP
Supplier:  Medchemexpress LLC HY1857050G
Encompass_Preferred
1-Naphthaleneacetic acid (1-Naphthylacetic acid), also known as NAA, is an auxin that can promote plant growth and induce root cluster formation. It is also an inhibitor of PLA2, with an IC50 of 13.16 μM. It is suitable for plant cell culture and is intended for research and analytical applications.
  • Promotes plant growth.
  • Induces root cluster formation.
  • Inhibitor of PLA2.
  • Suitable for plant cell culture.
  • Analytical standard available.
Catalog No. 50-003-87936