Learn More
Medchemexpress LLC 1-Naphthaleneboronic Acid (contains varying amounts of Anhydride) | 13922-41-3 | 100.0% | C10H9BO2 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHY78494
Encompass_Preferred
1-Naphthaleneboronic acid is a biochemical reagent useful as a biological material or organic compound for life science research. It is intended for research use only.
  • Appears as a white to off-white solid
  • Soluble in DMSO at 100 mg/mL (requires sonication)
  • Powder can be stored at -20°C for 3 years or 4°C for 2 years
  • Solutions can be stored at -80°C for 2 years or -20°C for 1 year
Catalog No. 50-003-57593