Learn More
Medchemexpress LLC 1-naphthalenecarboxaldehyde, 2-hydroxy- | 708-06-5 | MFCD00004005 | 99.9% | 172.18 g/mol | C11H8O2 | 50 G
SDP
Supplier:  Medchemexpress LLC HYW01688950G
Encompass_Preferred
CFL-137 is a small-molecule research compound described as a potent KRasG12C inhibitor with reported antiproliferative and anticancer activity; it is used in preclinical studies to investigate KRAS-driven cancers, including lung cancer. The compound has formula C11H8O2 and molecular weight 172.18 g/mol (CAS 708-06-5). Solubility in DMSO is ≥100 mg/mL. Store at 4°C under nitrogen; in solvent, store at -80°C (6 months) or -20°C (1 month).
  • Potent KRasG12C inhibition suitable for pathway studies.
  • Demonstrates antiproliferative and anticancer activity in preclinical models.
  • High solubility in DMSO (≥100 mg/mL) for assay preparation.
  • Stability guidelines provided for reliable sample handling.
  • Characterized chemical identity and reported high purity for reproducible experiments.
Catalog No. 50-003-02021