Learn More
Medchemexpress LLC 1-naphthalenemethanol | 4780-79-4 | MFCD00004044 | 100.0% | 158.20 g/mol | C11H10O | 1 ML

Supplier: Medchemexpress LLC HYW01724110MM/1ML
1-naphthalenemethanol (1-hydroxymethylnaphthalene) is a small organic compound used as a reagent and intermediate in organic synthesis and laboratory research. It is supplied as an off-white to yellow solid and may be purchased as a ready-to-use 10 mM solution in DMSO (1 mL). Refer to the safety data sheet for handling, storage, and hazard information.
- CAS number: 4780-79-4
- Molecular formula: C11H10O; molecular weight: 158.20 g/mol
- Appearance: off-white to yellow solid
- Available as solid in multiple gram quantities and as 10 mM solution in DMSO (1 mL)
- Analytical data: RP-HPLC report available; typical purity ~99.97%
- Applications: building block and intermediate in organic synthesis
- Storage and handling: consult SDS for safe use
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.