Learn More
Medchemexpress LLC 1-Naphthyl PP1 | 221243-82-9 | 99.4% | 317.39 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1394110MM/1ML
Encompass_Preferred
1-Naphthyl PP1 (1-NA-PP 1) is a selective inhibitor of src family kinases and Protein Kinase D. It has a molecular formula of C19H19N5, a molecular weight of 317.39, and a purity of 99.36% by HPLC. It inhibits v-Src and c-Fyn, c-Abl, CDK2 and CAMK II with IC50s of 1.0, 0.6, 0.6, 18 and 22 μM, respectively.
  • Selective inhibitor of src family kinases and Protein Kinase D.
  • Inhibits v-Src, c-Fyn, c-Abl, CDK2, and CAMK II.
Catalog No. 50-003-51075