Learn More
Medchemexpress LLC 1-naphthyl PP1 | 221243-82-9 | MFCD03425471 | 99.0% | 317.39 g/mol | C19H19N5 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY139415MG
Encompass_Preferred
1-Naphthyl PP1 is a small-molecule research inhibitor used to probe Src-family kinases and Protein Kinase D (PKD) signaling pathways in biochemical, cell-based, and in vivo studies.
  • Selective inhibitor of Src-family kinases and PKD.
  • Suitable for biochemical and cell-based kinase assays.
  • Formulations available as a solid or as a DMSO solution.
  • High purity (≈99%) with supporting analytical data.
  • Supplied with datasheet, COA, SDS, and handling instructions.
  • Used in studies of cell proliferation and kinase autophosphorylation.
Catalog No. 50-002-23225