Learn More
Selleck Chemical LLC 1-Naphthyl PP1 hydrochloride E2202-5MG
SDP
Supplier:  Selleck Chemical LLC E22025MG
Encompass_Preferred
1-Naphthyl PP1 hydrochloride (1-NA-PP 1 hydrochloride) is a selective inhibitor of Src family kinases inhibits v-Src c-Fyn c-Abl CDK2 and CAMKII with IC50s of 1 0 0 6 0 6 18 and 22 M respectively
Catalog No. 50-000-39258