Learn More
Medchemexpress LLC HY-13942 5mg Medchemexpress, 1-NM-PP1 CAS:221244-14-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY139425MG
Encompass_Preferred
Medchemexpress, HY-13942 5mg 1-NM-PP1 CAS:221244-14-0 1-NM-PP1, a cell-permeable PP1 analog, is a potent Src family kinases inhibitor with IC50s of 4.3 nM and 3.2 nM for v-Src-as1 and c-Fyn-as1, respectively[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0494
 Restricted Item