Learn More
Medchemexpress LLC 1-Nonadecanol | 1454-84-8 | 284.52 | 25 G

Supplier: Medchemexpress LLC 25GHYW004297
1-Nonadecanol is a white to off-white solid that is a component of supercritical carbon dioxide (SC-CO2) essential oil of *Heracleum thomsonii*. It is also recognized as an important aroma compound in *Neotinea ustulata*.
- Molecular formula: C19H40O
- Molecular weight: 284.52
- Purity: 99.2%
- Store powder at -20°C for up to 3 years or 4°C for up to 2 years
- Store in solvent at -80°C for up to 6 months or -20°C for up to 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.