Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-Nonadecanol | 1454-84-8 | 284.52 | 5 G
SDP

Supplier:  Medchemexpress LLC HYW0042975G

Encompass_Preferred

1-Nonadecanol is a chemical compound found as a component of supercritical carbon dioxide (SC-CO2) essential oil of Heracleum thomsonii and as an important aroma compound in Neotinea ustulata. It appears as a white to off-white solid.

  • White to off-white solid appearance
  • Purity of 99.2%
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month
  • Soluble in DMSO at 2 mg/mL (7.03 mM) with ultrasonic, warming, and heating to 60°C

Catalog No. 50-004-48907


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.