Learn More
Medchemexpress LLC 1-O-9Z-octadecenoyl-sn-glyceryl-3-phosphoric acid | 65528-98-5 | MFCD12912350 | 99.9% | 436.5 g/mol | C21H41O7P | 5 MG

Supplier: Medchemexpress LLC HY1378625MG
1-Oleoyl lysophosphatidic acid (1-oleoyl-LPA; CAS 65528-98-5) is a bioactive lysophospholipid that functions as a ligand for lysophosphatidic acid (LPA) receptors. It is used in cell biology and signaling research to activate LPA-mediated pathways and probe downstream effects such as SRE-driven transcription and β-galactosidase activity.
- Potent LPA receptor agonist used in signaling studies.
- High purity, typically 99.9% for research applications.
- Liquid form, colorless to light yellow.
- Molecular weight 436.5 g/mol and formula C21H41O7P.
- Soluble in ethanol for solution-based assays.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.