Learn More
Cayman Chemical 1-O-CaffeoylquInIc AcId 10mg
SDP
Supplier:  Cayman Chemical 3618910 MG
Encompass_Preferred
A phenolic acid; binds the NF-κB precursor protein p105 (Ki = 0.007 µM); binds PD-1 (KD = 12.4 µM) and inhibits the protein-protein interaction between PD-1 and PD-L1 (IC50 = 87.28 µM)
Catalog No. 50-291-2865