Learn More
Medchemexpress LLC 1-O-hexyl-2,3,5-trimethylhydroquinone | 148081-72-5 | 99.9% | 236.35 g/mol | C15H24O2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1007685MG
Encompass_Preferred
HTHQ (1-O-hexyl-2,3,5-trimethylhydroquinone) is a lipophilic phenolic antioxidant used in research to scavenge reactive oxygen species and study oxidative-stress related pathways. The compound is documented with CAS 148081-72-5, molecular weight 236.35 g/mol, and is supplied at 99.89% purity.
  • Lipophilic phenolic antioxidant that scavenges reactive oxygen species.
  • High purity, 99.89%, for reliable experimental results.
  • Suitable for in vitro and preclinical oxidative stress studies.
  • Small-quantity vials convenient for screening and dose-response work.
  • Documented quality and handling information available via COA and SDS.
Catalog No. 50-002-11327