Learn More
Medchemexpress LLC 1-O-trans-p-Coumaroylglycerol | 106055-11-2 | 95.51% | 238.24 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN97855MG
Encompass_Preferred
1-O-trans-p-Coumaroylglycerol is a compound found in certain plants with neuroprotective activity. It has shown potential in protecting against neurodegenerative diseases and is also believed to act as a natural antioxidant. Its applications may include improving cognitive function and supporting neurological health.
  • Neuroprotective activity
  • Potential in protecting against neurodegenerative diseases
  • Acts as a natural antioxidant
  • May enhance cognitive function
  • May support neurological health
Catalog No. 50-004-26027