Learn More
Medchemexpress LLC 1-octan-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-d17-ol | 153336-13-1 | 99.1% | 147.33 g/mol | C8HD17O | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW032013S100MG
Encompass_Preferred
1-Octanol-d17 is the deuterium-labeled analog of 1-octanol used as an isotopic/internal standard and tracer in analytical and research applications. The heptadeuterio (d17) isotopologue provides a predictable mass shift for accurate quantitation in mass spectrometry and NMR studies.
  • High isotopic labeling (d17) for unambiguous mass-shifted signals.
  • High purity (GC) suitable for analytical and quantitative work.
  • Available in small pack sizes for trace-level experiments.
  • Suitable as an internal standard in GC-MS and LC-MS workflows.
  • Stable when stored under recommended conditions; follow SDS for handling.
Catalog No. 50-003-00251