Learn More
Medchemexpress LLC 1-octanol-d2 | 78510-02-8 | 99.0% | 132.24 g/mol | C8H16D2O | 50 MG
SDP
Supplier:  Medchemexpress LLC HYW032013S150MG
Encompass_Preferred
1-Octanol-d2 is the deuterium-labeled analogue of 1-octanol provided as a small-pack stable-isotope reagent for analytical and research use. It serves as an internal standard or tracer in quantitative workflows and studies requiring isotopic labelling, and is suitable for NMR, GC-MS, and LC-MS applications.
  • Deuterium-labeled standard for analytical quantitation.
  • High isotopic enrichment (99.0%) for reliable mass shifts.
  • Compact 50 mg pack convenient for trace-level analyses.
  • Suitable as tracer and internal standard in NMR, GC-MS, and LC-MS.
  • Molecular weight 132.24 g/mol and formula C8H16D2O.
Catalog No. 50-002-99356