Learn More
Sigma Aldrich Fine Chemicals Biosciences 1-Octen-3-ol 98 fCC fG1KG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences W2805181KGK
Encompass
1-Octen-3-ol occurs naturally in mushrooms. It is the most prominent odor volatile produced by fungi such as Aspergillus Penicillium and fusarium species. 1-Octen-3-ol is also formed in major quantity when soybeans are soaked in water as a pretreatment before soymilk production. It is also a potent mosquito attractant. 1-Octen-3-ol is the major volatile compound formed during autoxidation of linoleic acid.
Catalog No. 50-289-9614