Learn More
Sigma Aldrich Fine Chemicals Biosciences 1 Octen 3 ol natural 95 fG100G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences W280523100GK
Encompass
1-Octen-3-ol occurs naturally in mushrooms. It is the most prominent odor volatile produced by fungi such as Aspergillus Penicillium and fusarium species. 1-Octen-3-ol is also formed in major quantity when soybeans are soaked in water as a pretreatment before soymilk production. It is also a potent mosquito attractant.
Catalog No. 50-290-2014