Learn More
Enzo Life Sciences 1-Oleoyl-2-acetyl-sn-glycerol, (50mg), CAS Number: 86390-77-4
SDP
Supplier:  Enzo Life Sciences BMLDG1250050
Encompass_Preferred
PKC activator. In vitro potent activator of protein kinase C (PKC). Stimulator of secretion and aggregation in blood platelets. Stimulates formation of actin nucleation sites. Stimulation of pp60c-src phosphorylation in EBV transformed B-cells, 470μM. Alternative Name: OAG. Formula: C23H42O5. MW: 398.6. CAS Number: 86390-77-4. Purity: ≥98% (TLC). Appearance: Colorless oil or waxy solid. Solubility: Soluble in DMSO or ethanol. Use/Stability: Stable for at least 1 year after receipt when stored, as supplied, at -20°C. Stock solutions are stable for up to 3 months at -20°C. Long Term Storage: -20°C
Catalog No. NC0170166