Learn More
Medchemexpress LLC 1-oleoyl-2-hydroxy-sn-glycero-3-PG sodium | 326495-24-3 | MFCD00674367 | 99.0% | 532.58 g·mol-1 | C24H46NaO9P | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1455055MG
Encompass_Preferred
1-Oleoyl-2-hydroxy-sn-glycero-3-PG sodium is a lysophospholipid (18:1 Lyso-PG) supplied as the sodium salt for biochemical research. It is provided with high purity for use in lipid biochemistry, membrane modeling, and signaling assays; handle and store per the SDS recommendations.
  • High purity (99.0%) suitable for biochemical assays.
  • Sodium salt form for improved solubility in aqueous buffers.
  • Provides an endogenous lysophospholipid for membrane and signaling studies.
  • Available in small, research-scale quantities for assay development.
  • Supplied with molecular weight and formula information for formulation work.
Catalog No. 50-002-21736