Learn More
Medchemexpress LLC 1-Oleoyl-sn-glycero-3-phosphocholine | 19420-56-5 | MFCD00133435 | 99.6% | 521.67 g/mol | C26H52NO7P | 50 MG
SDP
Supplier:  Medchemexpress LLC HYW14043950MG
Encompass_Preferred
1-Oleoyl-sn-glycero-3-phosphocholine is an unsaturated 18:1 lysophosphatidylcholine formed in plasma by lecithin-cholesterol acyltransferase (LCAT). It is supplied as a high-purity research standard for biochemical assays, lipidomics, and method development.
  • High purity (≈99.6%).
  • Molecular formula C26H52NO7P; molecular weight 521.67 g/mol.
  • Available in multiple pack sizes, including 50 mg.
  • Suitable for assay development, lipidomics, and biochemical studies.
  • Provided for research use only.
Catalog No. 50-002-82860