Learn More
Medchemexpress LLC 1-Palmitoyl-2-oleoyl-sn-glycero-3-PC (POPC) | 26853-31-6 | 99.9% | 100 MG
SDP
Supplier:  Medchemexpress LLC HY130462100MG
Encompass_Preferred
1-Palmitoyl-2-oleoyl-sn-glycero-3-PC (POPC) is a phospholipid and a major component of biological membranes. It is used for the preparation of liposomes and studying the properties of lipid bilayers.
  • Phospholipid
  • Major component of biological membranes
  • Used for liposome preparation
  • Used for studying lipid bilayers
  • Contains 16:0 and 18:1 fatty acids at sn-1 and sn-2 positions
  • White to off-white solid appearance
  • Structure classification: Lipid
  • Initial source: Plants, Microorganisms, widespread
Catalog No. 50-003-40425