Learn More
Hach Company Kit, 1 Bottle, 2.5 Gallon Polyethylene, for SD900 Portable Standard Sampler
Supplier:  Hach Company PS010030
Encompass
This kit provides a single 2.5 gallon polyethylene replacement bottle and the necessary hardware for use with SD900 portable standard samplers. It supplies a durable, chemical-resistant container and components to enable secure sample collection in field conditions.
  • Provides a 2.5 gallon polyethylene sample bottle for field collection.
  • Includes bottle support and a full-bottle shut-off for secure installation.
  • Polyethylene construction resists many chemicals and is lightweight for transport.
  • Single-bottle kit simplifies replacement and routine maintenance.
  • Designed for portable sampler installations and field sampling applications.
Catalog No. 50-002-07631