Learn More
Medchemexpress LLC 1-Pentadecanol | 629-76-5 | 228.41 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW00429510G
Encompass_Preferred
1-Pentadecanol is a long-chain fatty alcohol that exhibits anti-comedone activity and possesses moisturizing and hydrating properties, making it suitable for use in skin care products. It can also be utilized in the production of surfactants, emulsifiers, and lubricants.
  • Long-chain fatty alcohol
  • Exhibits anti-comedone activity
  • Possesses moisturizing and hydrating properties
  • Suitable for use in skin care products
  • Can be utilized in the production of surfactants
  • Can be utilized in the production of emulsifiers
  • Can be utilized in the production of lubricants
Catalog No. 50-004-45964