Learn More
Medchemexpress LLC 1-Pentanone, 1-(4-hydroxyphenyl)- | 2589-71-1 | 100.0% | 178.23 | C11H14O2 | 100 G
SDP
Supplier:  Medchemexpress LLC HYW016604100G
Encompass_Preferred
4'-Hydroxyvalerophenone is a small-molecule research reagent that inhibits 17β-hydroxysteroid dehydrogenase (17β-HSD). Supplied as a white to off-white solid, it is characterized by molecular formula C11H14O2 and molecular weight 178.23 g/mol. It is intended for laboratory research and biochemical screening applications.
  • High purity (99.96%) suitable for analytical and biological studies.
  • Inhibits 17β-hydroxysteroid dehydrogenase with IC50 ≈ 60.52 μM.
  • White to off-white solid, easy to weigh and handle.
  • Available in multiple pack sizes for small- to bulk-scale research.
Catalog No. 50-003-03776