Learn More
Medchemexpress LLC 1-Phenyl-2-pyrrolidi 10g
SDP
Supplier:  Medchemexpress LLC HYW10346310G
Encompass_Preferred
1-Phenyl-2-pyrrolidinone (1-Phenylpyrrolidin-2-one) is a phenyl analogue of GABA with sedative effect decreasing the exploratory behavior of rats at 50-100 mg/kg (i v ) 1-Phenyl-2-pyrrolidinone also has been proved to inhibit emotional reactions in dogs and cats 1-Phenyl-2-pyrrolidinone induces decreases in the pressor reaction to emotional stress without accompanied by normalization of the function of baroreceptor reflexes[1][2]
Catalog No. 50-002-06487