Learn More
Medchemexpress LLC 1-phenylpyrrole | 635-90-5 | MFCD00005343 | 99.9% | 143.19 g/mol | C10H9N | 10G

Supplier: Medchemexpress LLC HY12489110G
1-Phenylpyrrole is an aromatic heterocyclic compound used as a fluorescent probe and research reagent. It exhibits dual fluorescence-displaying a single emission band in nonpolar solvents and a red-shifted band in polar solvents-making it valuable for photophysical studies, solvent polarity sensing, and fluorescence spectroscopy.
- Dual fluorescence sensitive to solvent polarity.
- Suitable for fluorescence spectroscopy and photophysical research.
- High purity suitable for analytical and research applications.
- Available in gram-scale solid quantities for laboratory use.
- Soluble in DMSO at concentrations ≥ 2.5 mg/mL, forming a clear solution.
- Chemical formula C10H9N and molecular weight ~143.19 g/mol.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.