Learn More
Medchemexpress LLC 1-phenylsemicarbazide | 103-03-7 | MFCD00007942 | 97.0% | 151.17 g/mol | C7H9N3O | 50 G

Supplier: Medchemexpress LLC HYW28034950G
1-Phenylsemicarbazide is a small-molecule antifungal research chemical (CAS 103-03-7) commonly used in laboratory studies and in research on mold prevention for industrial materials. It is supplied as a solid with reported analytical purity and standard laboratory pack sizes. For research use only.
- Antifungal agent with reported activity against mold growth.
- Purity 97.0%.
- Molecular weight 151.17 g/mol.
- Chemical formula C7H9N3O.
- Available in common lab pack sizes, including 50 G.
- Supplied for research use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.