Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-Piperazinebutanol, α-(4-fluorophenyl)-4-(5-fluoro-2-pyrimidinyl)-, hydrochloride (1:1) | 105565-55-7 | 98.0% | 384.85 | 50 MG
SDP

Supplier:  Medchemexpress LLC HY10850910MG

Encompass_Preferred

BMY-14802 hydrochloride is a potent agonist of 5-HT1A and adrenergic α1 receptors, known for its antipsychotic effects.

  • Appearance: white to off-white solid.
  • Storage: -20°C, sealed storage, away from moisture. In solvent: -80°C, 6 months; -20°C, 1 month.
  • ¹H NMR spectrum consistent with structure.
  • MS consistent with structure.
  • Product tested and complies with given specifications.

Catalog No. 50-003-46537


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.