Learn More
Medchemexpress LLC 1-Piperidinecarboxylic acid, 3-hydroxy-, 1,1-dimethylethyl ester, (3S)- | 143900-44-1 | 98.9% | 201.27 | 50 G
SDP
Supplier:  Medchemexpress LLC HY4002350G
Encompass_Preferred
(S)-1-Boc-3-hydroxypiperidine is a chemical precursor that plays a crucial role in the synthesis of the anticancer agent Ibrutinib. This product is designed for research use and has been tested to comply with its specifications.
  • Key precursor for Ibrutinib synthesis.
  • Suitable for research use only.
  • Available in solid form.
  • Long-term stability with proper storage conditions.
Catalog No. 50-004-04507