Learn More
Medchemexpress LLC 1-piperidinecarboxylic acid, 3-hydroxy-, 1,1-dimethylethyl ester, (3S)- | 143900-44-1 | 98.94% | 201.27 | 25 G

Supplier: Medchemexpress LLC 25GHY40023
(S)-1-Boc-3-hydroxypiperidine is a key precursor in the synthesis of the anticancer agent Ibrutinib. This product is intended for research use only.
- Purity: 98.94%
- Appearance: Solid
- Color: White to off-white
- Storage: Powder at -20°C for 3 years or 4°C for 2 years; in solvent at -80°C for 6 months or -20°C for 1 month
- Solubility: Soluble in DMSO (100 mg/mL)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.