Learn More
Medchemexpress LLC 1-piperidinyloxy, 4-hydroxy-2,2,6,6-tetramethyl-d17,15N | 90429-66-6 | 98.4% | 190.34 | C9HD1715NO2 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY100561S1MG
Encompass_Preferred
Tempol-d17,15N is the deuterium- and 15N-labeled analog of Tempol, a superoxide dismutase (SOD)-mimetic aminoxyl. It is designed for use as a tracer or internal standard in analytical and mechanistic studies involving reactive oxygen species, and is suitable for quantitative analysis by NMR, GC-MS, and LC-MS.
  • Deuterium- and 15N-labeled for unambiguous tracer and internal standard use.
  • High purity (98.44%) suitable for analytical applications.
  • Compatible with NMR, GC-MS, and LC-MS workflows.
  • Useful in studies of reactive oxygen species and SOD-mimetic activity.
  • Supplied in small, low-mass quantities appropriate for tracer experiments.
Catalog No. 50-002-11501