Learn More
Medchemexpress LLC 1-Propanesulfonic acid, 3-(cyclohexylamino)- | 1135-40-6 | 100.0% | 100 G

Supplier: Medchemexpress LLC HYD0869100G
CAPS, also known as cyclohexylaminopropane sulfonic acid, is a surfactant that can be utilized as a biological buffer (0.05 M, pH 11) for dialysis. The CAPS surfactant carries a single negative charge as the amine is not ionized at high pH.
- Used as a biological buffer for dialysis
- Purity of 99.95%
- Molecular weight of 221.32
- Chemical formula C9H19NO3S
- Appears as a solid, white to off-white in color
- Soluble in H2O at 100 mg/mL and DMSO at 5 mg/mL
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.