Learn More
Medchemexpress LLC 1-Pyrenebutyric acid N-hydroxysuccinimide ester | 114932-60-4 | 99.93% | 385.41 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHYW013151
Encompass_Preferred
1-Pyrenebutyric acid N-hydroxysuccinimide ester (PANHS) is a linker and interface coupling agent used to prepare electrochemical biosensors. It can immobilize SARS-CoV-2 spike antibodies on graphene sheets. It is also commonly employed as an activation reagent for carboxylic acids in organic chemistry or biochemistry.
  • Linker and interface coupling agent
  • Immobilizes SARS-CoV-2 spike antibodies
  • Activation reagent for carboxylic acids
Catalog No. 50-004-51608