Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-pyrenesulfonic acid sodium | 59323-54-5 | MFCD00210088 | 99.4% | 304.30 g/mol | C16H9NaO3S | 100 MG
SDP

Supplier:  Medchemexpress LLC HYW127714100MG

Encompass_Preferred

1-Pyrenesulfonic acid sodium is the sodium salt of 1-pyrenesulfonic acid, a fluorescent organic reagent used in life-science research and analytical fluorescence applications. It is supplied as a solid with high purity and is suitable for use as a fluorescent probe, dye, or analytical standard.

  • High purity 99.4%.
  • Molecular weight 304.30 g/mol.
  • Chemical formula C16H9NaO3S.
  • Supplied as a solid for convenient handling and storage.
  • Suitable for fluorescence and analytical assays.
  • Available in small laboratory quantities such as 100 MG.

Catalog No. 50-002-83104


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.