Learn More
Medchemexpress LLC 1-pyrenesulfonic acid, sodium salt | 59323-54-5 | MFCD00210088 | 99.4% | 304.30 | C16H9NaO3S | 1 G
SDP
Supplier:  Medchemexpress LLC HYW1277141G
Encompass_Preferred
1-Pyrenesulfonic acid, sodium is the sodium salt of 1-pyrenesulfonic acid supplied as a solid research reagent for biochemical and analytical applications. It is an off-white to yellow solid with high purity (99.44% by HPLC), molecular weight 304.30 g/mol, and CAS 59323-54-5. Store sealed at 4°C; in solvent store -80°C (6 months) or -20°C (1 month).
  • High purity suitable for analytical and research use.
  • Solid form enables accurate weighing and dispensing.
  • Pyrene fluorophore useful for probe and labeling studies.
  • Stable when stored sealed under recommended temperatures.
  • Sodium salt provides aqueous solubility for buffer-based experiments.
Catalog No. 50-002-83105