Learn More
Medchemexpress LLC 1-Pyrrolidine-2,2,3,3,4,4-d6-acetamide, α-ethyl-5-oxo-, (αS)- | 1133229-30-7 | 98.7% | 176.25 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY1102251MG
Encompass_Preferred
Levetiracetam-d6 (ring-d6) is an internal standard designed for determining Levetiracetam content in whole blood. This product is for research use only and not sold to patients.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-44756